NMR tubes with large writing area, high throughput, 5 mm Ext.Ø., SP Wilmad-LabGlass
Leverandør: SP Industries
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WG-1003-7 WG-1003-7-25 WG-1003-7-50
634-1178EA 1460 DKK
634-1178 634-1179 634-1180
NMR tubes with large writing area, high throughput, 5 mm Ext.Ø., SP Wilmad-LabGlass
Rør NMR rør
Double the writing area allows for easy sample marking and identification.
  • 2× larger marking area eliminates cramped and unreadable sample names
  • Silk screened writing area keeps sample names from easily wiping off
  • Designed for routine use in most low to mid field NMR spectrometers
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimensions
These high throughput tubes have an average camber of 60 µm to ensure spectrum clarity for smallMore Product Information