PfuTurbo DNA polymerase
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
600250 600252 600254 600256 600322 600324 600410 600412 600414 600600 600602
AGLS600250EA 1460 DKK
AGLS600250 AGLS600252 AGLS600254 AGLS600256 AGLS600322 AGLS600324 AGLS600410 AGLS600412 AGLS600414 AGLS600600 AGLS600602
PfuTurbo DNA polymerase
Nukleinsyrereagenser End-point PCR Enzymes and Kits
This DNA polymerase is ideal for a variety of techniques requiring high-fidelity DNA synthesis by the polymerase chain reaction (PCR).
PfuTurbo DNA Polymerase is an enhanced version of Pfu DNA Polymerase for robust, high-fidelity PCR. PfuTurbo DNA polymerase is a mixture of cloned Pfu DNA polymerase and the exclusive thermostable ArchaeMaxx polymerase-enhancing factor that enhances PCR product yields and increases target length capability without altering DNA replication fidelity.
PfuTurbo DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuTurbo DNA Polymerase for robust, high-fidelity PCR.
PfuTurbo Hotstart DNA Polymerase is a special formulation of high fidelity PfuTurbo DNA Polymerase with hot start activity.
PfuTurbo Cx Hotstart DNA Polymerase is a specially formulated with a mutant of Pfu DNA Polymerase designed to overcome uracil sensitivity for more robust performance.
The PfuTurbo Hotstart PCR Master Mix is a 2X formulation of PfuTurbo hotstart DNA Polymerase, an optimised PCR reaction buffer, magnesium, and dNTPs.