Brilliant II SYBR® Master mix
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
600825 600826 600828 600831 600834
AGLS600825EA 7853 DKK
AGLS600825 AGLS600826 AGLS600828 AGLS600831 AGLS600834
Brilliant II SYBR® Master mix
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
Brilliant II SYBR® QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
  • Earlier Ct detection
  • Improved data reproducibility
  • Greater flexibility for use with numerous different templates and targets
  • More accurate quantification results
  • Developed on the Agilent Mx QPCR systems
When compared to other commercially available QPCR reagents, we observe earlier threshold cycle (Ct) detection at high and low template concentrations as well as improved reaction efficiency and replicate reproducibility across the entire range of template concentrations.
Advarsel: For research use only. Not for use in diagnostic procedures.