Hybrid automated microscope (upright + inverted + fluorescence + transmitted light), ECHO Revolution
Leverandør: DISCOVER ECHO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VIBE0002 RNCU0001 RNJY0001 RNSDEW51 RNCO2001 RNCU2001 RNHM2001
ECHLVIBE0002EA 27810 DKK
ECHLVIBE0002 630-3340 630-3341 630-3342 630-3354 630-3355 630-3356
Hybrid automated microscope (upright + inverted + fluorescence + transmitted light), ECHO Revolution
Mikroskoper
This 'all in one' automated hybrid microscope can be used for upright and inverted observation methods.
  • Live cell imaging - view and capture live samples with automated time-lapse microscopy
  • Stage top incubator (optional) - ensures optimal conditions for maintaining specimen viability
  • Multi-channel - capture and overlay multiple wavelengths in fluorescence
  • Multi-point - set multiple acquisition points to view and revisit over time
  • Image stitching - capture and stitch to form large FOV high resolution images
  • Z-stack - acquire and stack images at multiple focal planes
  • Auto focus - automatically find and track best Z–plane(s) for focus
  • Hyperscan - high speed image stitching
5 MP monochrome camera and 5 MP Brightfield CMOS Color Camera. Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used. Hyperscan Software (optional) - ultra fast scanning and mosaic image stitching at high-speed camera frame rates.
Illumination technique: Brightfield and Epi-fluorescence (phase contrast available as an option). Ultra-bright LEDs with 50000-hour lifespan. Touch screen functions drive cameras and illumination, replacing manual turrets and levers. Traditional eyepieces are replaced by a 32" 4K display for easier viewing. The upright observation method is ideal for viewing glass slides. Useful for water dipping applications. The inverted observation method is suitable for viewing live samples in fluidic chambers such as dishes and well plates. It also allows for easier manipulation of samples. Ideal for long working distance applications.
Bestillingsoplysninger: To be completed with objectives, filters and light cubes, and condensers (all sold separately).
Leveringsinformation:
Supplied with a 32" 4K monitor, keyboard and mouse, motorised epi-fluorescence turret, brightfield and fluorescence cameras, power supply, automated precision X/Y stage, and lens cloth.