Tris Buffers, Sterile
Leverandør: G BiosciencesSamlet vurdering
Gennemsntilig vurdering
Tris is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane, with the formula (HOCH2)3CNH2. Tris is extensively used in as a major component in buffer solutions.
- Sterile
- Ready to use