Thermomyces lanuginosa lipase
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIIM1002-100G BIIM1002-25G
APOSBIIM1002-100GEA 4200 DKK
APOSBIIM1002-100G APOSBIIM1002-25G
Thermomyces lanuginosa lipase
Enzymer
Covalent on IB-150A. Activity: 10000 TBU/g, 5000 PLU/g
  • immobilized on Immobead 150