Cathepsin/subtilisin inhibitor
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIC2003-1MG BIC2003-5MG
APOSBIC2003-1MGEA 1210 DKK
APOSBIC2003-1MG APOSBIC2003-5MG
Cathepsin/subtilisin inhibitor
Proteiner
Selectively inhibits members of Cystein protease family including Cathepsin L and members of Serine protease family including Subtilisin, Carlsberg and Thermitase