BHD Autoclavable Muff Covers, with Elastic
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFNWB04 MFNWB05 MFNWB06 MFNWB07 MFNWB08 MFNWB09 MFNWB12 MFNWB16 MFNWB18 MFNWB18SK02 MFNWB18SK02TL MFNWB05-SP MFNWB08-SP MFNWB12-SP MFNWB03
129-1760EA 16340 DKK
129-1760 129-1761 129-1762 129-1763 129-1764 129-1765 129-1766 129-1767 129-1768 129-2474 129-2475 129-3059 129-3062 129-3064 129-3058
BHD Autoclavable Muff Covers, with Elastic
Covers
Save time and standardise activities in equipment preparation for sterilisation.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with sheets and tape
BHD muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
Some items feature a 2" (51 mm) extension skirt, to allow removal of the muff without inadvertently touching the critical surfaces.
Leveringsinformation: Certificates of Conformance sent with each shipment.
Emballering: Conveniently packed in 100 per sealed poly bag.