Polypropylene Muff Covers with Elastic
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFNWHD04 MFNWHD05 MFNWHD06 MFNWHD07 MFNWHD08 MFNWHD09 MFNWHD12 MFNWHD16 MFNWHD18
129-3067EA 16340 DKK
129-3067 129-3068 129-3069 129-3070 129-3071 129-3072 129-3073 129-3074 129-3075
Polypropylene Muff Covers with Elastic
Covers
Keeps cleaned non sterile production hoses protected from external sources of contamination.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear and puncture resistant
  • Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Leveringsinformation: Certificates of Conformance sent with each shipment.