Fluoride CRM, Standard Solutions, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.32233.0100 1.32234.0100 1.32235.0100 1.32236.0100
1.32233.0100EA 722 DKK
1.32233.0100 1.32234.0100 1.32235.0100 1.32236.0100
Fluoride CRM, Standard Solutions, Supelco®
Standarder Anioniske standarder Fluoride Standards
NaF in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certificeringer: Traceable to SRM from NIST and USP.
Bestil nu
Specifikationer
Environmentally Preferable
M-Clarity quality level MQ200