2,3,4-Trifluoro-5-methylbenzoyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC303440-1G PC303440-5G
APOSPC303440-1GEA 1570 DKK
APOSPC303440-1G APOSPC303440-5G
2,3,4-Trifluoro-5-methylbenzoyl chloride
2,3,4-Trifluoro-5-methylbenzoyl chloride
Formel: C₈H₄ClF₃O
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1003709-50-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR