Exhaust filters for barrels, b.safe
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M506-21 M506-23 M506-25
590-0179EA 2770 DKK
590-0179 590-0180 590-0181
Exhaust filters for barrels, b.safe
Filtre Ventilationsfiltre
Filter body made of PE with connection thread for direct installation on barrels. Lid with aeration openings and label field to monitor service life. Filling made of activated carbon for adsorption of solvents.
  • Activation of service life through touch of the button and/or individual marking on the back side
  • Splash-proof ventilation openings
  • Activated carbon in the inside guarantee adsorption of harmful vapours
  • PP for high resistance and fire class rating V-0
  • Plenty of space for fittings and hose connectors
Safe disposal of solvents without leakage of harmful solvent vapours. The activated carbon in the b.safe exhaust filter restrains solvent vapours from the escaping air during filling. The service life of the filter can be easily monitored through documentation of the exchange date by hand in the label field on the lid upon installation. We recommend the following filter sizes: Size XL for 60 to 100 L, size XXL for barrels having 100 L and more.