VWR®, Precision Grip Gloves, NBR Coating
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112-5740EA 241 DKK
112-5740 112-5737 112-5735 112-5736 112-5739 112-5738
VWR®, Precision Grip Gloves, NBR Coating
Handsker
This VWR® precision grip glove (with nitrile coating) provides basic protection against mechanical risks.
  • Nylon composition
  • Nitrile (NBR) coating on palm and fingertips
  • Blue knitted glove with black coating
Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment. Also great for package handling, picking and packing.
Certificeringer: Meets requirements for EN 388:2016 (4131X). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.