2,5-Difluorobenzhydrazide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC9952-1G PC9952-5G
APOSPC9952-1GEA 785 DKK
APOSPC9952-1G APOSPC9952-5G
2,5-Difluorobenzhydrazide
2,5-Difluorobenzhydrazide
Formel: C₇H₆F₂N₂O
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02183537
CAS-nummer: 265644-03-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR