Bismuth(III)trifluormethansulphonat
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Bismuth triflate, Bismuth(III) trifluoromethanesulphonate
PC9890-100G
PC9890-25G
PC9890-5G
APOSPC9890-100GEA
1830
DKK
APOSPC9890-100G
APOSPC9890-25G
APOSPC9890-5G
Bismuth(III)trifluormethansulphonat
Bismuth(III)trifluormethansulphonat
Formel:
C₃BiF₉O₉S₃ Smeltepunkt: >300 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02093669 CAS-nummer: 88189-03-1 UN nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...