4-Bromo-2,3,5,6-tetrafluoropyridine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC9701-1G
PC9701-5G
APOSPC9701-1GEA
436
DKK
APOSPC9701-1G
APOSPC9701-5G
4-Bromo-2,3,5,6-tetrafluoropyridine 95%
4-Bromo-2,3,5,6-tetrafluoropyridine
Formel:
C₅BrF₄N Kogepunkt: 134…135 °C Massefylde: 1,92 g/cm³ (25 °C) Flammepunkt: 63 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00075169 CAS-nummer: 3511-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...