1-Brom-2-Chlor-3-Fluorbenzen
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-3-fluorobromobenzene
PC9674-25G
PC9674-5G
PC9674-1G
PC9674-10G
APOSPC9674-25GEA
275
DKK
APOSPC9674-25G
APOSPC9674-5G
APOSPC9674-1G
APOSPC9674-10G
1-Brom-2-Chlor-3-Fluorbenzen
1-Brom-2-Chlor-3-Fluorbenzen
Formel:
C₆H₃BrClF Molekylvægt: 209.44 g/mol Smeltepunkt: 29 °C Massefylde: 1.7 g/cm³ (25 °C) Flammepunkt: 78 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04038306 CAS-nummer: 883499-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...