Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC9443-1G PC9443-500MG PC9443-5G
APOSPC9443-1GEA 1160 DKK
APOSPC9443-1G APOSPC9443-500MG APOSPC9443-5G
Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate 97%
Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate
Formel: C₁₄H₁₁F₃O₃S
Molekylvægt: 316.3 g/mol
Smeltepunkt: 60…61 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00202083
CAS-nummer: 343375-98-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR