[1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Methyl-5-(phenylthio)-3-(trifluoromethyl)-1H-pyrazol-4-ylmethanol

PC9436-1G PC9436-500MG PC9436-5G
APOSPC9436-1GEA 4000 DKK
APOSPC9436-1G APOSPC9436-500MG APOSPC9436-5G
[1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
[1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Formel: C₁₂H₁₁F₃N₂OS
Smeltepunkt: 45…46 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00172634
CAS-nummer: 318239-51-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR