3-Fluor-2-formylpyridin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC9249-1G PC9249-25G PC9249-5G
APOSPC9249-1GEA 183 DKK
APOSPC9249-1G APOSPC9249-25G APOSPC9249-5G
3-Fluor-2-formylpyridin 97%
3-Fluor-2-formylpyridin
Formel: C₆H₄FNO
Smeltepunkt: 51…54 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07781234
CAS-nummer: 31224-43-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR