5-Fluoro-2-methylbenzotrifluoride 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Fluoro-1-methyl-2-(trifluoromethyl)benzene
PC907074-1G
PC907074-25G
PC907074-5G
APOSPC907074-1GEA
208
DKK
APOSPC907074-1G
APOSPC907074-25G
APOSPC907074-5G
5-Fluoro-2-methylbenzotrifluoride 99%
5-Fluoro-2-methylbenzotrifluoride
Formel:
C₈H₆F₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11226544 CAS-nummer: 141872-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...