5-Bromo-3-fluoro-2-methylpyridine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC906562-250MG PC906562-1G PC906562-5G
APOSPC906562-250MGEA 590 DKK
APOSPC906562-250MG APOSPC906562-1G APOSPC906562-5G
5-Bromo-3-fluoro-2-methylpyridine 95%
5-Bromo-3-fluoro-2-methylpyridine
Formel: C₆H₅BrFN
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12196979
CAS-nummer: 1162674-74-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR