5,8-Difluoroquinoline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC901859-1G PC901859-5G
APOSPC901859-1GEA 2180 DKK
APOSPC901859-1G APOSPC901859-5G
5,8-Difluoroquinoline 98%
5,8-Difluoroquinoline
Formel: C₉H₅F₂N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01939053
CAS-nummer: 16650-32-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR