4-Methyl-2-nitro-1-(trifluoromethyl)benzene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Nitro-4-(trifluoromethyl)toluene

PC901215-1G PC901215-250MG PC901215-5G
APOSPC901215-1GEA 436 DKK
APOSPC901215-1G APOSPC901215-250MG APOSPC901215-5G
4-Methyl-2-nitro-1-(trifluoromethyl)benzene 95%
4-Methyl-2-nitro-1-(trifluoromethyl)benzene
Formel: C₈H₆F₃NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16140172
CAS-nummer: 154057-13-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR