3,4-Difluoro-2-methoxyaniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC8768-1G PC8768-5G
APOSPC8768-1GEA 589 DKK
APOSPC8768-1G APOSPC8768-5G
3,4-Difluoro-2-methoxyaniline
3,4-Difluoro-2-methoxyaniline
Formel: C₇H₇F₂NO
Molekylvægt: 159.133 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07368889
CAS-nummer: 114076-35-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR