2-Bromo-3-fluorophenol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC8606-1G PC8606-25G PC8606-5G
APOSPC8606-1GEA 164 DKK
APOSPC8606-1G APOSPC8606-25G APOSPC8606-5G
2-Bromo-3-fluorophenol 98%
2-Bromo-3-fluorophenol
Formel: C₆H₄BrFO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04037220
CAS-nummer: 443-81-2
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR