2-Fluor-4-pyridinboronsyrepinacolester
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PC8520-1G
PC8520-5G
APOSPC8520-1GEA
218
DKK
APOSPC8520-1G
APOSPC8520-5G
2-Fluor-4-pyridinboronsyrepinacolester
2-Fluor-4-pyridinboronsyrepinacolester
Formel:
C₁₁H₁₅BFNO₂ Molekylvægt: 223.05 g/mol Smeltepunkt: 47…51 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06798253 CAS-nummer: 458532-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...