5-Fluor-2-methoxyphenol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Fluor-2-hydroxyanisole

PC8212-100G PC8212-25G PC8212-5G
APOSPC8212-100GEA 2220 DKK
APOSPC8212-100G APOSPC8212-25G APOSPC8212-5G
5-Fluor-2-methoxyphenol 98%
5-Fluor-2-methoxyphenol
Formel: C₇H₇FO₂
Molekylvægt: 142.13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00143459
CAS-nummer: 72955-97-6
UN nummer: 3265
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR