4-Fluor-2-methoxybenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC8209-25G PC8209-5G PC8209-100G
APOSPC8209-25GEA 709 DKK
APOSPC8209-25G APOSPC8209-5G APOSPC8209-100G
4-Fluor-2-methoxybenzoesyre
4-Fluor-2-methoxybenzoesyre
Formel: C₈H₇FO₃
Molekylvægt: 170.14 g/mol
Smeltepunkt: 134…136 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00673010
CAS-nummer: 395-82-4

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR