3-Chlor-5-fluorphenol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC8165-1G PC8165-5G PC8165-25G
APOSPC8165-1GEA 164 DKK
APOSPC8165-1G APOSPC8165-5G APOSPC8165-25G
3-Chlor-5-fluorphenol 98%
3-Chlor-5-fluorphenol
Formel: C₆H₄ClFO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042212
CAS-nummer: 202982-70-5
UN nummer: 3265
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR