Zinkfluoramid, anhydrous 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC8030-100G
PC8030-250G
PC8030-25G
APOSPC8030-100GEA
680
DKK
APOSPC8030-100G
APOSPC8030-250G
APOSPC8030-25G
Zinkfluoramid, anhydrous 99%
Zinkfluoramid
Formel:
F₂Zn Kogepunkt: 1500 °C Smeltepunkt: 872 °C Massefylde: 4,95 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011298 CAS-nummer: 7783-49-5 EINECS: 232-001-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...