3-Chlor-5-(trifluormethyl)phenol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-Chloro-5-hydroxybenzotrifluoride
PC8004-1G
PC8004-25G
PC8004-5G
APOSPC8004-1GEA
546
DKK
APOSPC8004-1G
APOSPC8004-25G
APOSPC8004-5G
3-Chlor-5-(trifluormethyl)phenol
3-Chlor-5-(trifluormethyl)phenol
Formel:
C₇H₄ClF₃O Molekylvægt: 196.56 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04115852 CAS-nummer: 570391-18-3 UN nummer: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...