2-Bromo-N-tert-butyl-5-fluorobenzamide 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC7911-1G
PC7911-25G
PC7911-5G
APOSPC7911-1GEA
765
DKK
APOSPC7911-1G
APOSPC7911-25G
APOSPC7911-5G
2-Bromo-N-tert-butyl-5-fluorobenzamide 98%
2-Bromo-N-tert-butyl-5-fluorobenzamide
Formel:
C₁₁H₁₃BrFNO Smeltepunkt: 116…118 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09475888 CAS-nummer: 941294-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...