(2-Fluoro-5-nitrophenyl)acetic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Fluoro-5-nitrophenylacetic acid

PC7840-10G PC7840-25G PC7840-100G
APOSPC7840-10GEA 284 DKK
APOSPC7840-10G APOSPC7840-25G APOSPC7840-100G
(2-Fluoro-5-nitrophenyl)acetic acid
(2-Fluoro-5-nitrophenyl)acetic acid
Formel: C₈H₆FNO₄
Smeltepunkt: 149…150 °C
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04091773
CAS-nummer: 195609-18-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR