3-(Trifluoromethyl)cyclohexanone 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC7668M-1G
PC7668M-5G
APOSPC7668M-1GEA
752
DKK
APOSPC7668M-1G
APOSPC7668M-5G
3-(Trifluoromethyl)cyclohexanone 97%
3-(Trifluoromethyl)cyclohexanone
Formel:
C₇H₉F₃O Kogepunkt: 177…179 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00034662 CAS-nummer: 585-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...