2-(Trifluoromethyl)benzhydrol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Phenyl(2-(trifluoromethyl)phenyl)methanol

PC7512-25G PC7512-5G
APOSPC7512-25GEA 4560 DKK
APOSPC7512-25G APOSPC7512-5G
2-(Trifluoromethyl)benzhydrol 97%
2-(Trifluoromethyl)benzhydrol
Formel: C₁₄H₁₁F₃O
Kogepunkt: 273…274 °C
Massefylde: 1,311 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014396
CAS-nummer: 727-98-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR