3-Bromo-5-(trifluoromethyl)benzoic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC7295-25G
PC7295-5G
APOSPC7295-25GEA
240
DKK
APOSPC7295-25G
APOSPC7295-5G
3-Bromo-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₄BrF₃O₂ Smeltepunkt: 133…135 °C Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03412186 CAS-nummer: 328-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...