2-Chlor-3,6-difluorbenzaldehyd
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC7284-5G
PC7284-25G
PC7284-100G
APOSPC7284-5GEA
218
DKK
APOSPC7284-5G
APOSPC7284-25G
APOSPC7284-100G
2-Chlor-3,6-difluorbenzaldehyd
2-Chloro-3,6-difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Smeltepunkt: 46…50 °C Flammepunkt: 110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01631443 CAS-nummer: 261762-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...