Ethyl (2-fluorophenyl)acetate 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl 2-fluorophenylacetate
PC7118-25G
PC7118-5G
APOSPC7118-25GEA
881
DKK
APOSPC7118-25G
APOSPC7118-5G
Ethyl (2-fluorophenyl)acetate 99%
Ethyl (2-fluorophenyl)acetate
Formel:
C₁₀H₁₁FO₂ Kogepunkt: 235 °C Massefylde: 1,101 g/cm³ (25 °C) Flammepunkt: 85 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04039343 CAS-nummer: 584-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...