3-fluordipicolinsyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC7057-25G PC7057-5G PC7057-1G
APOSPC7057-25GEA 610 DKK
APOSPC7057-25G APOSPC7057-5G APOSPC7057-1G
3-fluordipicolinsyre
3-fluordipicolinsyre
Formel: C₆H₄FNO₂
Smeltepunkt: 167…169 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04114108
CAS-nummer: 152126-31-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR