Perfluorobutane-1-sulphonyl chloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Nonafluorobutane-1-sulphonyl chloride

PC6996-1G PC6996-5G PC6996-25G
APOSPC6996-1GEA 709 DKK
APOSPC6996-1G APOSPC6996-5G APOSPC6996-25G
Perfluorobutane-1-sulphonyl chloride 97%
Perfluorobutane-1-sulphonyl chloride
Formel: C₄ClF₉O₂S
Kogepunkt: 100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03412262
CAS-nummer: 2991-84-6
UN nummer: 3265
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR