p-Toluenesulphonyl fluoride 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: p-toluenesulphonylfluoride

PC6991-25G PC6991-100G
APOSPC6991-25GEA 828 DKK
APOSPC6991-25G APOSPC6991-100G
p-Toluenesulphonyl fluoride 98%
p-Toluenesulphonyl fluoride
Formel: C₇H₇FO₂S
Kogepunkt: 112 °C/16mmHg
Smeltepunkt: 40…41 °C
Flammepunkt: 105 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007421
CAS-nummer: 455-16-3
UN nummer: 3261
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR