2-Amino-3-methylbenzotrifluoride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methyl-6-(trifluoromethyl)aniline

PC6989-10G PC6989-1G PC6989-5G
APOSPC6989-10GEA 3720 DKK
APOSPC6989-10G APOSPC6989-1G APOSPC6989-5G
2-Amino-3-methylbenzotrifluoride
2-Amino-3-methylbenzotrifluoride
Formel: C₈H₈F₃N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03701069
CAS-nummer: 88301-98-8
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR