3-Bromo-4-fluoropyridine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC6885-1G PC6885-5G PC6885-10G
APOSPC6885-1GEA 164 DKK
APOSPC6885-1G APOSPC6885-5G APOSPC6885-10G
3-Bromo-4-fluoropyridine 98%
3-Bromo-4-fluoropyridine
Formel: C₅H₃BrFN
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04114101
CAS-nummer: 116922-60-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR