Lithium-bis(trifluormethylsulfonyl)imid 99%
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt
PC6834-50G PC6834-5G PC6834-25G PC6834-100G
APOSPC6834-50GEA 436 DKK
APOSPC6834-50G APOSPC6834-5G APOSPC6834-25G APOSPC6834-100G
Lithium-bis(trifluormethylsulfonyl)imid 99%
Lithium-bis(trifluormethylsulfonyl)imid
Formel: C₂F₆LiNO₄S₂
Molekylvægt: 287.09 g/mol
Smeltepunkt: 234…238 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00210017
CAS-nummer: 90076-65-6
EINECS: 415-300-0
UN nummer: 2923
ADR: 8+6.1 II