3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC6764-25G
PC6764-5G
APOSPC6764-25GEA
5560
DKK
APOSPC6764-25G
APOSPC6764-5G
3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde 95%
3-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde
Formel:
C₉H₆F₄O₂ Kogepunkt: 95…97 °C/9mmHg Massefylde: 1,393 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
CAS-nummer:
35295-35-3 EINECS: 252-496-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...