6-(Trifluoromethyl)pyridine-3-amidoxime 94%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-(Trifluoromethyl)nicotinamidoxime
PC6696-1G
PC6696-5G
APOSPC6696-1GEA
600
DKK
APOSPC6696-1G
APOSPC6696-5G
6-(Trifluoromethyl)pyridine-3-amidoxime 94%
6-(Trifluoromethyl)pyridine-3-amidoxime
Formel:
C₇H₆F₃N₃O Smeltepunkt: 192…195 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06245538 CAS-nummer: 386704-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...