Fomblin® Z-25

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Perfluoropolyether YR-1800

PC6484-25G PC6484-5G PC6484-5KG
APOSPC6484-25GEA 0 DKK
APOSPC6484-25G APOSPC6484-5G APOSPC6484-5KG
Fomblin® Z-25
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized

A linear PFPE available as a neutral or functionalized fluid, used in aerospace, magnetic media and polymer modification uses.

Kogepunkt: >270 °C
Smeltepunkt: –75 °C
Massefylde: 1.85 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00163699
CAS-nummer: 69991-67-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR