Natriumhexafluorantimonat 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Sodium hexafluoroantimonate(V)
PC6480-1KG
PC6480-250G
PC6480-25G
PC6480-100G
PC6480-500G
PC6480-5KG
APOSPC6480-1KGEA
1530
DKK
APOSPC6480-1KG
APOSPC6480-250G
APOSPC6480-25G
APOSPC6480-100G
APOSPC6480-500G
APOSPC6480-5KG
Natriumhexafluorantimonat 98%
Natriumhexafluorantimonat
Formel:
F₆NaSb Smeltepunkt: 1360 °C Massefylde: 3,375 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003482 CAS-nummer: 16925-25-0 EINECS: 240-989-8 UN nummer: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...