Natriumchlordifluoracetat 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC6460-100G
PC6460-25G
PC6460-500G
APOSPC6460-100GEA
436
DKK
APOSPC6460-100G
APOSPC6460-25G
APOSPC6460-500G
Natriumchlordifluoracetat 97%
Natriumchlordifluoracetat
Formel:
C₂ClF₂NaO₂ Smeltepunkt: 195…198 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064771 CAS-nummer: 1895-39-2 EINECS: 217-586-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...